Learn More
Waters Corp Waters OBD Prep Column, CSH C18, 10 μm, 10 x 100 mm
Supplier:  Waters Corp 186008269
Encompass_Preferred
CSH C18 columns are general-purpose columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology helps to address common issues such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Alternative selectivity compared to other C18 sorbents
  • Charged Surface Hybrid (CSH) technology helps overcome column-related problems
  • Addresses peak shape asymmetry for basic compounds
  • Improves loading for basic compounds
  • Reduces column bleed
  • Facilitates faster mobile-phase re-equilibration
Catalog No. NC1779606