Learn More
Waters Corp Waters Prep Guard Cartridge, CSH C18, 130Å, 5 μm, 19 mm X 10 mm
Supplier:  Waters Corp 186005418
Encompass_Preferred
This prep guard cartridge uses Charged Surface Hybrid (CSH) technology to address common chromatography challenges. It offers diverse analyte selectivity and unmatched reproducibility. Made from the same packing material as analytical columns, it ensures compatibility and extends column lifetime, reducing purification and lab costs.
  • Utilizes CSH technology to address peak shape asymmetry, poor loading, and column bleed.
  • Provides diverse analyte selectivity and unmatched reproducibility.
  • Ensures compatibility and extended analytical column lifetime.
  • Offers superior peak shape for basic compounds and high peak capacity.
  • Increases mass load for preparative separation without ion-pairing reagents.
  • Enables rapid column equilibration and quick changes to mobile-phase conditions.
  • Reduces purification and lab costs.
Catalog No. NC0781458