Learn More
Waters Corp Waters XSelect CSH C18 Method Validation Kit, 130 Å, 2.5 μm, 4.6 × 100 mm
Supplier:  Waters Corp 186006238
Encompass_Preferred
This method validation kit comprises three batches of columns, each packed with a different batch of sorbent, ensuring comprehensive method validation.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 11
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • 130 Å pore size
  • Surface area of 185
  • Compatible with UPLC and UHPLC systems
  • CSH particle technology
  • L1 USP classification
  • 4.6 mm inner diameter
  • 100 mm length
  • 15% carbon load
  • Three units per package
Catalog No. 50-201-589