Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Column, 2.5 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 176002893
Encompass_Preferred
This XSelect CSH Fluoro-Phenyl Column supports reversed-phase separation with low pH mobile phases. Its non-encapped sorbent enhances acidic compound retention, while ultra-low dispersion hardware ensures exceptional HPLC assay performance.
  • Maximizes selectivity differences and method development freedom with low pH mobile phases.
  • Non-encapped sorbent provides enhanced retention of acidic compounds.
  • 2.5 μm XP columns offer exceptional separation performance, robustness, and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.
  • Manufactured under stringent cGMP and ISO 9001 standards, ensuring high quality.
Catalog No. 50-465-933