Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Column, 130Å, 3.5 μm, 4.6 mm × 150 mm
Supplier: Waters Corp 186005325RF
This XSelect CSH Fluoro-Phenyl Column has a pore size of 130Å, particle size of 3.5 μm, and dimensions of 4.6 mm × 150 mm. It comes with an eConnect Column Tag.
- Contains unique product-specific identifying information, such as part and serial numbers.
- Offers convenient access to product information via NFC technology.
- Automatically imports data to HPLC system software.
- Simplifies setup and reduces manual data entry with pre-equipped tag.
- Permanently affixed tag reduces compliance concerns and aids auto-detectability.
- Tag design enhances detection and mitigates interference in temperature-sensitive methods.
- Ensures identical quality standards and performance as untagged counterparts.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.