Learn More
Waters Corp XSelect CSH Fluoro-Phenyl Column, 130Å, 5 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186005337
Encompass_Preferred
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-791-363