Learn More
Waters Corp Waters XSelect CSH Fluoro-Phenyl Column, 5 µm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186005330
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl column is engineered to maximize selectivity differences, offering method development flexibility, especially with low pH mobile phases. Its non-encapped sorbent design provides enhanced retention for acidic compounds.
  • Maximizes selectivity differences for diverse analytical needs.
  • Offers maximum method development freedom, particularly with low pH mobile phases.
  • Provides enhanced retention of acidic compounds due to non-encapped sorbent.
Catalog No. 50-791-356