Learn More
Waters Corp Waters Column, XSelect CSH Fluoro-Phenyl, 5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186005329
Encompass_Preferred
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides enhanced retention of acidic compounds.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 1 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Non-endcapped sorbent
  • Fluoro-Phenyl bonding technology
  • Low silanol activity
  • Spherical particle shape
  • 5 μm particle size
  • 130 Å pore size
  • 185 surface area
  • Suitable for HPLC systems
  • CSH particle technology
  • USP classification L43
  • 2.1 mm inner diameter
  • 50 mm length
  • 10% carbon load
  • One unit per package
Catalog No. 50-791-355