Learn More
Waters Corp Waters Column, CSH Fluoro-Phenyl, 3.0 x 75 mm, 2.5 μm, 130 Å
Supplier:  Waters Corp 186006118
Encompass_Preferred
This column provides exceptional separation performance, robustness, and throughput for HPLC, UHPLC, and UPLC Technology platforms. It offers enhanced selectivity for various compounds and is designed for high-pressure applications.
  • Withstands pressures up to 18,000 psi
  • Offers exceptional separation performance and robustness for HPLC assays
  • Compatible with HPLC, UHPLC, and UPLC Technology platforms
  • Directly scalable to 1.7 μm, 1.8 μm, 3.5 μm, and 5 μm columns
  • Fluorinated phases provide alternative selectivity to standard C18 phases
  • Enhanced selectivity for acidic, alkaline, and neutral compounds
  • Superior peak shape for basic compounds and excellent batch-to-batch reproducibility
Catalog No. 50-179-527