Learn More
Waters Corp Waters Column, PFP/Fluoro-Phenyl, 130Å, 2.5 μm, 2.1 mm x 75 mm
Supplier:  Waters Corp 186006114
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl column is designed to maximize selectivity differences, offering method development freedom, especially with low pH mobile phases. Its non-encapped sorbent provides enhanced retention of acidic compounds. The column is packed in ultra-low dispersion hardware, and the 2.5 μm eXtended Performance [XP] columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC adoption.
  • Maximizes selectivity differences
  • Enhanced retention of acidic compounds
  • Ultra-low dispersion hardware
  • Exceptional separation performance, robustness, and throughput for HPLC assays
  • Seamless transition path toward UPLC adoption
Catalog No. 50-179-523