Learn More
Waters Corp Waters Column, CSH Fluoro-Phenyl, 130Å, 2.5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186006113
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl column maximizes selectivity, offering method development freedom, especially with low pH mobile phases. Its non-encapped sorbent enhances retention of acidic compounds. These 2.5 μm XP columns, packed in ultra-low dispersion hardware, enable exceptional separation, robustness, and throughput for HPLC assays, providing a transition to UPLC. All column documents are available online, reducing paper waste.
  • Maximizes selectivity; method development freedom for low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • 2.5 μm XP columns for exceptional performance.
  • Ultra-low dispersion hardware for superior separation.
  • Exceptional separation, robustness, and throughput for HPLC assays.
  • Transition to UPLC adoption.
  • Online documents for easy access and reduced waste.
Catalog No. 50-179-522