Learn More
Waters Corp Waters Corporation Column, Fluoro-Phenyl, 130Å, 2.5 μm, 3 x 50 mm
Supplier: Waters Corp 186006117
This XSelect CSH Fluoro-Phenyl column is designed to maximize selectivity differences, offering maximum method development freedom, especially with low pH mobile phases. The non-encapped sorbent enhances retention of acidic compounds. It is packed in ultra-low dispersion hardware, and its 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a seamless transition to UPLC adoption.
- PFP/Fluoro-Phenyl chemistry.
- Reversed phase separation mode.
- Hybrid particle substrate.
- pH range from 1 to 8.
- Maximum pressure of 18000 psi (1240 Bar).
- Low silanol activity.
- Particle size of 2.5 μm.
- Pore size of 130 Å.
- Inner diameter of 3 mm and length of 50 mm.
- Compatible with UPLC and UHPLC systems.
- USP classification L43.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.