Learn More
Waters Corp Waters Corporation XSelect CSH Fluoro-Phenyl Column, 130Å, 2.5 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186006115
Encompass_Preferred
The XSelect CSH Fluoro-Phenyl column maximizes selectivity differences and method development freedom, especially with low pH mobile phases. Its non-encapped sorbent enhances retention of acidic compounds. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns offer exceptional separation, robustness, and throughput for HPLC assays, enabling seamless UPLC transition.
  • Maximize selectivity differences with low pH mobile phases.
  • Enhanced retention of acidic compounds (non-encapped sorbent).
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm XP columns provide exceptional separation, robustness, and throughput for HPLC.
  • Seamless transition to UPLC adoption.
  • Delivers high-quality, accurate results in liquid chromatography.
  • Built in specialized facilities, adhering to cGMP and ISO 9001 standards.
Catalog No. 50-179-524