Learn More
Waters Corp XSelect CSH Fluoro-Phenyl XP Column, 130Å, 2.5 µm, 2.1 mm X 75 mm, 3/pk
Supplier:  Waters Corp 176002604
Encompass_Preferred
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-263