Learn More
Waters Corp Waters Column, Fluoro-Phenyl, 2.5 μm, 130Å, 3 mm x 30 mm
Supplier:  Waters Corp 186006116
Encompass_Preferred
This CSH Fluoro-Phenyl column maximizes selectivity differences, offering method development freedom, especially with low pH mobile phases. Its non-encapped sorbent enhances retention of acidic compounds. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns ensure exceptional separation performance, robustness, and throughput for HPLC assays, enabling a seamless transition to UPLC adoption.
  • Maximizes selectivity differences.
  • Offers method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
  • Certificate of Analysis documents available for download.
Catalog No. 50-179-525