Learn More
Waters Corp XSelect Method Validation Kit, 130Å, 2.5 μm, 3 mm × 150 mm, 3/pk
Supplier:  Waters Corp 186006792
Encompass_Preferred
This method validation kit contains three XSelect CSH Phenyl-Hexyl columns with varying sorbent batches. They offer alternative analyte selectivity, crucial for method development. The robust C6 phenyl-ligand selectively retains polyaromatic compounds via pi-pi interactions. These 2.5 μm XP columns ensure exceptional separation and throughput for HPLC assays, supporting UPLC adoption.
  • Three columns with different sorbent batches for method validation.
  • Alternative analyte selectivity for method development.
  • Robust, low-bleed C6 phenyl-ligand.
  • Selectively retains polyaromatic compounds via pi-pi interactions.
  • 2.5 μm XP columns in ultra-low dispersion hardware for exceptional separation.
  • Facilitates seamless transition to UPLC.
Catalog No. 50-466-484