Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Column, 130 Å, 3.5 μm
Supplier:  Waters Corp 186005380RF
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development. They feature a robust, low bleed C6 phenyl-ligand, selectively retaining polyaromatic compounds through pi-pi interactions.
  • Offers alternative analyte selectivity
  • Valuable tool for method development
  • Trifunctionally bonded C6 phenyl-ligand
  • Robust, low bleed sorbent
  • Selectively retains polyaromatic compounds via pi-pi interactions
  • Equipped with an electronic eConnect column tag
  • NFC-enabled for automatic identification, tracking, and digital tracing of column use history
  • Compatible with any HPLC system
  • Advantages realized with Alliance iS System running Empower software
Catalog No. 50-239-0906