Learn More
Waters Corp Waters Column, CSH Phenyl-Hexyl, 5 μm, 4.6 mm, 150 mm
Supplier:  Waters Corp 186005400
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity for method development, using a robust C6 phenyl-ligand for selective polyaromatic compound retention. They offer superior performance, comparable backpressure, and enhance LC in acidic, low ionic strength mobile phases.
  • Alternative selectivity for method development.
  • Robust C6 phenyl-ligand for polyaromatic retention.
  • Superior performance and comparable backpressure.
  • Increased loading capacity and particle size scalability.
  • CSH technology enhances LC.
  • Optimized selectivity with low pH mobile phases.
  • Non-end-capped sorbent enhances acidic chemical retention.
  • Ultra-low dispersion hardware for separation performance.
  • Smooth transition to UPLC.
Catalog No. 50-791-417