Learn More
Waters Corp Waters XSelect CSH Phenyl-Hexyl Column, 130Å, 3.5 μm, 1 mm X 150 mm
Supplier:  Waters Corp 186005361
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide you with alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • Alternative analyte selectivity
  • Trifunctionally bonded C6 phenyl-ligand
  • Robust, low bleed sorbent
  • Selectively retains polyaromatic compounds through pi-pi interactions
Catalog No. 50-791-380