Learn More
Waters Corp Waters Corporation Column, Phenyl-Hexyl, 130Å, 3.5 μm, 4.6 mm X 150 mm
Supplier:  Waters Corp 186005380
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions. Based on novel Charged Surface Hybrid (CSH) technology, these columns are designed to improve virtually all elements of LC column performance in acidic, low ionic strength mobile phases commonly used by chromatographers.
  • Provides alternative analyte selectivity compared to traditional reversed-phase columns.
  • Trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent.
  • Offers superior peak shape for basic compounds, increased loading capacity, and excellent stability at low pH.
  • Ensures improved reproducibility between batches of material.
Catalog No. 50-203-7703