Learn More
Waters Corp XSelect Method Validation Kit, Phenyl-Hexyl, 130Å, 3.5 μm
Supplier:  Waters Corp 186005563
Encompass_Preferred
These HPLC columns, powered by CSH Technology, offer wide selectivities and superior performance in acidic, low ionic strength, LC/MS friendly mobile phases. This Method Validation Kit includes three columns, each packed with a distinct gel lot of stationary phase, providing an alternative analyte selectivity for method development.
  • Compatible with all HPLC, UPLC, and UHPLC technologies.
  • Offers superior separation performance, resilience, and throughput.
  • Provides improved performance and backpressure compared to core-shell columns.
  • Features higher loading capacity and greater particle size scalability.
  • Charged Surface Hybrid (CSH) technology enhances liquid chromatography performance.
  • Provides alternate analyte selectivity to conventional reversed-phase columns.
  • Non-end-capped sorbent enhances retention of acidic compounds.
Catalog No. 50-791-520