Learn More
Waters Corp Waters Corporation Column, 2.5 μm, 130 Å, 2.1 mm × 100 mm
Supplier:  Waters Corp 186006127
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns offer an alternative analyte selectivity compared to traditional reversed-phase columns, making them a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition to UPLC adoption.
  • All column documents, including Certificates of Analysis, are available digitally, eliminating paper COAs and eCords.
Catalog No. 50-179-532