Learn More
Waters Corp Waters Corporation Column, XSelect CSH Phenyl-Hexyl, 130Å, 2.5 μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186006135
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns offer alternative analyte selectivity compared to traditional reversed-phase columns, making them a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through π–π interactions.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.
Catalog No. 50-201-549