Learn More
Waters Corp XSelect CSH Phenyl-Hexyl XP Column, 130Å, 2.5 µm, 3 mm X 50 mm, 3/pk
Supplier:  Waters Corp 176002615
Encompass_Preferred
XSelect CSH Phenyl-Hexyl columns provide you with alternative analyte selectivity compared to traditional reversed-phase columns and are a valuable tool for method development. The trifunctionally bonded C6 phenyl-ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-274