Learn More
Waters Corp XSelect CSH Phenyl-Hexyl Method Validation Kit, 130Å, 2.5 µm, 4.6 mm X 50 mm, 3/pk
Supplier:  Waters Corp 186006249
Encompass_Preferred
This method validation kit consists of three batches of XSelect CSH Phenyl-Hexyl columns packed with three different batches of sorbent
Catalog No. 50-201-592