Learn More
Waters Corp Waters Column, C18, 2.5 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 186006736
Encompass_Preferred
This column features general purpose, high performance C18 chemistry, offering exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, its 2.5 μm eXtended Performance [XP] technology enables superior separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC.
  • Superior peak shape for basic chemicals.
  • Enhanced retention and outstanding column lifetimes at low pH.
  • Minimum column bleed and great batch-to-batch consistency.
  • Ultra-efficiencies and seamless column format conversion.
  • High Strength Silica (HSS) technology for improved polar component retention.
  • HSS particles provide mechanical strength for enduring UPLC pressures.
  • Completely transferable to UPLC separations and scalable.
Catalog No. 50-466-444