Learn More
Waters Corp XSelect Method Validation Kit, C18, 3.5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186006410
Encompass_Preferred
The HSS C18 packing is the most stable commercially available bonded phase. It does not suffer from peak shape issues that affect older stationary phases relying on steric protection and lack of endcapping for stability at low pH. The HSS C18 HPLC columns are fully scalable and completely transferable to UPLC separations.
  • Most stable commercially available bonded phase.
  • Does not suffer from peak shape issues associated with older stationary phases.
  • Fully scalable and transferable to UPLC separations.
  • Endcapped.
  • Low silanol activity.
  • Spherical particle shape.
  • Designed for HPLC systems.
Catalog No. 50-201-615