Learn More
Waters Corp Waters Column, C18, 2.1 mm x 150 mm, 2.5 μm, 100 Å
Supplier:  Waters Corp 186006742
Encompass_Preferred
The XSelect HSS C18 SB column is a low coverage, unendcapped C18 phase designed for low pH separations of complex basic and non-basic compounds. It offers alternative analyte selectivity and exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC.
  • Low coverage, unendcapped C18 phase for low pH separations
  • Offers alternative analyte selectivity and exceptional separation performance
  • Provides robustness and high throughput for HPLC assays
  • Facilitates seamless transition to UPLC adoption
  • Developed with HSS—High Strength Silica technology
  • Withstands UPLC pressures for scalable and transferable UPLC separations
  • Delivers different selectivity and improved separation of polar compounds
Catalog No. 50-466-450