Learn More
Waters Corp Waters Corporation XSelect HSS C18 Column, 3.5 μm, 4.6 mm x 75 mm
Supplier:  Waters Corp 186006428
Encompass_Preferred
XSelect HPLC Columns offer diverse selectivity, high analyte loading, operational stability, and full scalability for UPLC®, HPLC, and preparative separation platforms. High Strength Silica (HSS) Technology increases polar compound retention and offers different selectivity than more hydrophobic BEH and CSH particles.
  • Designed for wide range of chromatographic separations.
  • Offers diverse selectivity and high analyte loading.
  • Provides operational stability.
  • Scalable for UPLC®, HPLC, and preparative platforms.
  • HSS Technology for increased polar compound retention.
  • Delivers different selectivity than hydrophobic BEH and CSH particles.
  • Mechanically strong to tolerate UPLC pressures.
  • Seamlessly scalable with equivalent 1.8 μm HSS particles.
Catalog No. 50-179-693