Learn More
Waters Corp Waters XSelect HSS C18 SB XP Column, 2.5 μm, 2.1 mm x 75 mm
Supplier:  Waters Corp 186006162
Encompass_Preferred
The XSelect HSS C18 SB column is a low coverage, unendcapped C18 phase designed for low pH separations of complex mixtures. It provides alternative analyte selectivity compared to modern, high coverage, fully endcapped C18 stationary phases. These 2.5 μm eXtended Performance (XP) columns are packed in ultra-low dispersion hardware, enabling exceptional separation performance, robustness, and throughput for HPLC assays, and offer a seamless transition path toward UPLC adoption.
  • Low coverage, unendcapped C18 phase for low pH separations of complex mixtures of basic and non-basic compounds.
  • Offers alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases.
  • Packed in ultra-low dispersion hardware (XP columns) for exceptional separation performance, robustness, and throughput in HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-179-554