Learn More
Waters Corp Waters XSelect HSS C18 SB XP Column, 2.5 μm, 2.1 mm ID
Supplier:  Waters Corp 186006163
Encompass_Preferred
The XSelect HSS C18 SB column is a low coverage, unendcapped C18 phase for low pH separations of complex mixtures. These 2.5 μm XP columns provide exceptional separation performance, robustness, and throughput for HPLC assays, facilitating transition to UPLC.
  • Improved peak shape for basic compounds and minimal column bleed.
  • Good batch-to-batch consistency and ultra-efficiencies.
  • Easy transferability between column formats and fully scalable for UPLC separations.
  • High Strength Silica (HSS) technology enhances polar compound retention and selectivity.
  • Improves resolution of complex samples; offers enhanced selectivity for various compounds.
  • Offers improved retention for basic polar compounds in low-pH mobile phases.
  • Operates at high pressures, up to 18000 psi (1240 Bar).
  • Compatible with a VanGuard Cartridge for extended lifetime and performance.
Catalog No. 50-179-555