Learn More
Waters Corp HPLC Column, XSelect HSS C18 SB, 2.5 μm, 3 × 100 mm
Supplier: Waters Corp 186006167
This C18 high performance chemistry is universally applicable, offering excellent retention and peak shape. These “eXtended Performance” columns use ultra-low dispersion hardware for exceptional separation, robustness, and high throughput in HPLC assays, allowing a seamless transition to UPLC. They are designed for reproducible and selective chromatographic separations, utilizing Charged Surface Hybrid (CSH) and High Strength Silica (HSS) particle technologies.
- “eXtended Performance” [XP] columns
- Ultra-low dispersion hardware
- Exceptional separation, robustness, and high throughput for HPLC assays
- Seamless transition to UPLC
- Inner diameter: 3 mm
- Length: 100 mm
- Particle size: 2.5 μm
- Carbon load: 8.0%
- Endcapping: No
- Mode: Reversed Phase (RP)
- pH range: 2 to 8
- Pore size: 100 Å
- USP number: L1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.