Learn More
Waters Corp Waters Column, C18, 2.5 μm, 100 Å, 2.1 mm × 75 mm
Supplier:  Waters Corp 176002636
Encompass_Preferred
The HSS C18 SB column is a low coverage, unendcapped C18 phase, suitable for low pH separations involving complex mixtures of basic and non-basic compounds. It offers an alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance (XP) columns provide exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-179-295