Learn More
Waters Corp WATERS CORPORATION Analytical Column, 2.5 μm, 2.1 x 50 mm, HSS C18
Supplier: Waters Corp 176002619
This XSelect Analytical Column is designed for high-performance liquid chromatography, offering exceptional separation performance, robustness, and throughput for various assays. It features 2.5 μm particles and is compatible with HPLC, UHPLC, and UPLC Technology platforms, engineered to withstand high pressures.
- 2.5 μm particle size for high-resolution separations.
- Dimensions: 2.1 x 50 mm for optimal column efficiency.
- Constructed from High Strength Silica (HSS) material.
- Trifunctional C18 ligand type for versatile chromatography.
- Features a pH range of 1 to 8.
- Engineered for enhanced retention of basic polar compounds with low-pH mobile phases.
- Designed to withstand pressures up to 18,000 psi.
- Provides increased polar compound retention and different selectivity due to 100% silica HSS particle.
- Quantity: 3 per pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.