Learn More
Waters Corp Waters Column, XSelect HSS C18, 100Å 2.5 μm, 4.6 x 50 mm
Supplier: Waters Corp 186006145
XSelect HSS C18 is a general-purpose, high-performance C18 chemistry with exceptional retention and peak shape for all compounds. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
- C18 chemistry for exceptional retention and peak shape.
- Packed in ultra-low dispersion hardware.
- 2.5 μm eXtended Performance (XP) columns for outstanding separation.
- Supports HPLC assays and UPLC adoption.
- Separation mode: Reversed Phase.
- Particle substrate: Silica.
- pH range: 1 to 8.
- Maximum pressure: 10000 psi (690 bar).
- Particle size: 2.5 μm.
- Pore size: 100 Å.
- Dimensions: 4.6 mm inner diameter x 50 mm length.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.