Learn More
Waters Corp Waters HPLC Column, Cyano (CN), 3.5 μm, 2.1 mm, 100 mm
Supplier:  Waters Corp 186005909
Encompass_Preferred
The propyl cyano bonded phase of this column offers ultra-performance as a general purpose chemistry. It provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH.
  • Provides superior peak shape for bases.
  • Offers increased retention.
  • Delivers extremely long column lifetimes at low pH.
  • Fully scalable and transferable to ACQUITY UPLC separations.
Catalog No. 50-179-382