Learn More
Waters Corp Waters XSelect HSS Cyano (CN) Column, 3.5 μm, 2.1 mm × 150 mm, 100Å
Supplier:  Waters Corp 186005910
Encompass_Preferred
The XSelect HSS Cyano (CN) Column features an ultra-performance general-purpose propyl cyano bonded phase. It delivers superior peak shape for bases, increased retention, and extended column lifetimes at low pH. Supporting both reversed-phase and normal-phase chromatography, it utilizes HSS technology for enhanced analytical performance.
  • Fully scalable and transferable to ACQUITY UPLC separations.
  • Enables both reversed-phase and normal-phase chromatography.
  • Uses high strength silica (HSS) technology for enhanced chromatographic performance and different selectivity, especially for polar molecules.
  • Offers mechanical strength for UPLC pressures.
  • Cost-effective for reducing retention of heavily retained compounds by allowing less organic solvent in the mobile phase, speeding up chromatographic analysis.
Catalog No. 50-179-383