Learn More
Waters Corp XSelect HSS Cyano (CN) Column, 100Å, 3.5 µm, 2.1 mm X 30 mm, 1/pk
Supplier:  Waters Corp 186005906
Encompass_Preferred
The XSelect HSS CN chemistry is an ultra-performance general purpose propyl cyano bonded phase that provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH. XSelect HSS CN HPLC columns are fully scalable and transferable to ACQUITY UPLC separations.
Catalog No. 50-179-379