Learn More
Waters Corp Waters HPLC Column, XSelect HSS Cyano, 3.5 μm, 4.6 mm, 50 mm, 100 Å, pH 2–8
Supplier:  Waters Corp 186005919
Encompass_Preferred
This XSelect HSS Cyano column offers low hydrophobicity for rapid elution and alternative selectivity. It provides exceptional performance for HPLC assays, enabling seamless UPLC transition. With High Strength Silica (HSS) technology, it ensures reproducible and highly selective chromatographic separations.
  • Highly stable cyano phase with low hydrophobicity.
  • Offers alternative selectivity to conventional alkyl chain columns.
  • Designed for reproducible and highly selective chromatographic separations.
  • Utilizes High Strength Silica (HSS) particle technology.
  • Can be used for both normal–phase and reversed–phase separations.
  • Mechanically strong to tolerate UPLC pressures.
  • Ensures superior peak shape and excellent batch–to–batch reproducibility.
Catalog No. 50-179-392