Learn More
Waters Corp Waters Corporation Cyano Column, 100Å, 5 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 186005932
Encompass_Preferred
The chemistry is an ultra-performance general-purpose propyl cyano bonded phase designed to provide superior peak shape for bases, increased retention, and extended column lifetimes at low pH. These columns are fully scalable and transferable to ACQUITY UPLC separations.
  • Ultra-performance general-purpose propyl cyano bonded phase
  • Superior peak shape for bases
  • Increased retention
  • Extremely long column lifetimes at low pH
  • Fully scalable and transferable to UPLC separations
Catalog No. 50-179-404