Learn More
Waters Corp XSelect Column, Cyano (CN), 5 μm, 4.6 x 150 mm
Supplier:  Waters Corp 186005945
Encompass_Preferred
The XSelect HSS CN chemistry is an ultra-performance general purpose propyl cyano bonded phase that provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH. These columns are fully scalable and transferable to ACQUITY UPLC separations.
  • Ultra-performance general purpose propyl cyano bonded phase.
  • Provides superior peak shape for bases.
  • Increased retention.
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to ACQUITY UPLC separations.
Catalog No. 50-179-416