Learn More
Waters Corp Method Validation Kit, HSS CN, 5 μm, 3.0 x 100 mm
Supplier:  Waters Corp 186005956
Encompass_Preferred
This Method Validation Kit includes 3 columns, each from a different batch. These HPLC columns offer high analyte loading, operational stability, and full scalability for UPLC, HPLC, and preparative separation platforms. They feature superior peak shape for basic compounds, low column bleed, excellent batch-to-batch reproducibility, and ultra-efficiencies.
  • Exceptional selectivity achieved through carefully chosen ligands and proprietary High Strength Silica (HSS) technology.
  • Versatile HSS CN phase for successful use in both normal-phase and reversed-phase separations.
  • Three columns from different batches for robust method validation studies.
  • Seamlessly scalable with chemical and physical characteristics matching 1.8 μm ACQUITY UPLC HSS particles.
  • Excellent batch-to-batch reproducibility and transferability.
Catalog No. 50-179-427