Learn More
Waters Corp Waters XSelect HSS PFP XP Column, 2.5 μm, 3.0 x 75 mm
Supplier: Waters Corp 176002648
The XSelect HSS PFP column is a 2.5 μm XP column, offering outstanding separation performance for HPLC assays and seamless transition to UPLC.
- Low coverage, unendcapped PFP phase.
- Unique selectivity for closely related compounds.
- Ultra-low dispersion hardware.
- High throughput, robust for HPLC.
- Seamless UPLC transition.
- PFP/Fluoro-Phenyl chemistry; reversed phase; silica substrate.
- pH 2 to 8; Max pressure: 18000 psi (1240 Bar).
- Particle size: 2.5 μm; Pore size: 100 Å; 7% carbon load.
- 3 mm ID, 75 mm length.
- Multi-system compatibility (UPLC, UHPLC, HPLC).
- Scalable to 1.7 μm to 5 μm columns.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.