Learn More
Waters Corp XSelect HSS PFP Method Validation Kit, 100Å, 2.5 µm, 4.6 mm X 50 mm, 3/pk
Supplier:  Waters Corp 186006273
Encompass_Preferred
This method validation kit consists of three batches of XSelect HSS PFP columns packed with three different batches of sorbent
Catalog No. 50-201-600