Learn More
Waters Corp Waters XSelect Column, 2.5 μm, 3 x 30 mm, 100 Å
Supplier: Waters Corp 176002646
The XSelect HSS PFP column is a low coverage, unendcapped pentafluorophenyl phase, offering exceptional separation of closely related compounds and alternate selectivity to traditional alkyl chain columns. Its intricate retention mechanisms and ultra-low dispersion hardware, along with 2.5 μm eXtended Performance [XP] columns, ensure robust, high-throughput HPLC assays and a seamless transition to UPLC.
- PFP/Fluoro-Phenyl chemistry
- Reverse phase separation mode
- Silica particulate substrate
- pH range of 2 to 8
- Maximum pressure of 18000 psi (1240 bar)
- Spherical particle shape
- Particle diameter of 2.5 μm
- Pore dimensions of 100 Å
- Internal diameter of 3 mm
- Length of 30 mm
- Equipped with an eCord
- Suitable for UPLC and UHPLC systems
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.