Learn More
Waters Corp Waters Column, HSS PFP, 100Å, 2.5 μm, 3 mm x 30 mm
Supplier: Waters Corp 186006176
The XSelect HSS PFP column provides exceptional separation of closely related compounds with alternate selectivity to traditional alkyl chain columns. These 2.5 μm eXtended Performance [XP] columns offer outstanding separation performance, robustness, and throughput for HPLC assays, enabling a smooth transition to UPLC.
- Chemistry: PFP/Fluoro-phenyle
- Separation mode: Reversed-phase
- Support particles: Silica
- pH range: 2 to 8
- Maximum pressure: 18,000 psi (1,240 bar)
- Not endcapped
- Grafting technology: PFP
- Silanol activity: High
- Particle shape: Spherical
- Particle size: 2.5 μm
- Endfitting type: Parker-style
- Pore size: 100 Å
- System: UPLC, UHPLC
- Particle technology: HSS
- Internal diameter: 3 mm
- Length: 30 mm
- Carbon load: 7%
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.