Learn More
Waters Corp XSelect HSS T3 XP Column, 100Å, 2.5 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186006740
Encompass_Preferred
The XSelect HSS T3 is a low-coverage C18 phase compatible with 100% aqueous mobile phase and should be your first choice when developing separations for polar and non-polar compounds. The XSelect HSS T3 columns exhibit ultra-low MS bleed and are fully scalable and transferable to UPLC separations. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-448