Learn More
Waters Corp Waters HPLC Column, 100Å, 5 μm, 3 mm x 30 mm
Supplier:  Waters Corp 186006477
Encompass_Preferred
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
  • Compatible with 100% aqueous mobile phase.
  • Provides ultra-low MS bleed.
  • Promotes superior polar compound retention.
  • Fully scalable and transferable to UPLC separations.
Catalog No. 50-179-731