Learn More
Waters Corp XSelect HSS T3 Column, 100 Å, 5 μm, 4.6 mm X 150 mm
Supplier:  Waters Corp 186004791RF
Encompass_Preferred
The HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column for retaining extremely polar compounds by reversed-phase in high aqueous mobile phases. It features an electronic eConnect column tag with unique column-specific information, enabling automatic identification, tracking, and digital tracing of column use history. Advantages are unlocked with a compatible system running Empower software.
  • Fully endcapped, low-coverage C18 bonded-phase column designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases.
  • Equipped with an electronic eConnect column tag containing unique column-specific information.
  • NFC-enabled device-tagged columns developed to automatically identify, track and digitally trace column use history.
  • Can be used on any HPLC, but its advantages are unlocked when used with a compatible system.
Catalog No. 50-239-1103