Learn More
Waters Corp XSelect HSS T3 Method Validation Kit, 100Å, 3.5 µm, 2.1 mm X 100 mm, 3/pk
Supplier:  Waters Corp 186006488
Encompass_Preferred
The trifunctional alkyl C18 bonded phase used for the HSS T3 sorbents is compatible with 100% aqueous mobile phase and provides ultra-low MS bleed, while promoting superior polar compound retention. HSS T3 HPLC columns are fully scalable and transferable to UPLC separations.
Catalog No. 50-201-633